siRNA Details
| Gene Symbol: LYSMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286812.1 | NP_001273741.1 | 3 | 1195 | ||
| NM_198273.2 | NP_938014.1 | 3 | 1254 | |||
| XM_005248421.4 | XP_005248478.1 | 3 | 1719 | |||
| GenBank mRNA | AK291478.1 | - | 3 | 1256 | ||
| AL137315.1 | - | 1 | 756 | |||
| AL832721.1 | - | 3 | 1235 | |||
| BC058027.1 | - | 3 | 1196 | |||
| BC136740.1 | - | 3 | 1151 | 9020730 | ||
| BC136741.1 | - | 3 | 1146 | 9020731 | ||
| BX537972.1 | - | 3 | 1229 | |||
| CR933624.1 | - | 4 | 845 | |||

LYSMD3
Genomic Map