siRNA Details
| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080435.2 | NP_001073904.1 | 6 | 1460 | ||
| XM_005272421.4 | XP_005272478.1 | 6 | 1076 | |||
| XM_005272422.3 | XP_005272479.1 | 6 | 823 | |||
| XM_005272423.4 | XP_005272480.1 | 6 | 1062 | |||
| XM_011521232.2 | XP_011519534.1 | 4 | 1272 | |||
| XM_017021923.1 | XP_016877412.1 | 6 | 2909 | |||
| GenBank mRNA | AB075851.1 | - | 6 | 1446 | ||
| AK126887.1 | - | 6 | 1461 | |||

WHAMM
Genomic Map