siRNA Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080824.2 | NP_001074293.1 | 5 | 1191 | ||
| NM_001277053.1 | NP_001263982.1 | 6 | 1338 | |||
| XM_011532504.1 | XP_011530806.1 | 6 | 1138 | |||
| GenBank mRNA | AK128325.1 | - | 3 | 2230 | ||
| AK296608.1 | - | 6 | 1175 | |||
| BC051789.1 | - | 5 | 1192 | |||
| JQ653415.1 | - | 6 | 1132 | |||

TRABD2A
Genomic Map