siRNA Details
| Gene Symbol: TMEM120B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080825.2 | NP_001074294.2 | 1 | 191 | ||
| XM_017018853.1 | XP_016874342.1 | 1 | 191 | |||
| XM_017018854.1 | XP_016874343.1 | 1 | 191 | |||
| GenBank mRNA | AK126568.1 | - | 1 | 191 | ||
| BC127768.1 | - | 1 | 69 | 40127741 | ||
| BC127769.1 | - | 1 | 69 | 40127743 | ||

TMEM120B
Genomic Map