siRNA Details
| Gene Symbol: LINC00934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024246.2 | 4 | 1476 | |||
| GenBank mRNA | AK057497.1 | - | 4 | 1476 | ||
| BC101457.1 | - | 1 | 479 | |||
| BC101458.1 | - | 1 | 479 | |||
| BC101459.1 | - | 1 | 479 | |||
| BC118554.1 | - | 1 | 479 | |||
| BC118632.1 | - | 2 | 393 | |||

LINC00934
Genomic Map