siRNA Details
| Gene Symbol: FRMD6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037676.1 | 1 | 1912 | |||
| GenBank mRNA | AK022437.1 | - | 1 | 1913 | ||
| BX104826.1 | - | 1 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRMD6-AS1| Gene Symbol: FRMD6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037676.1 | 1 | 1912 | |||
| GenBank mRNA | AK022437.1 | - | 1 | 1913 | ||
| BX104826.1 | - | 1 | 270 | |||