siRNA Details
| Gene Symbol: LOC146795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135222.1 | 3, 4 | 1738 | |||
| GenBank mRNA | AK057377.1 | - | 3, 4 | 1610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC146795| Gene Symbol: LOC146795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135222.1 | 3, 4 | 1738 | |||
| GenBank mRNA | AK057377.1 | - | 3, 4 | 1610 | ||