siRNA Details
| Gene Symbol: SMIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193628.1 | NP_001180557.1 | 4 | 872 | ||
| XM_017026303.1 | XP_016881792.1 | 5 | 1895 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM17| Gene Symbol: SMIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193628.1 | NP_001180557.1 | 4 | 872 | ||
| XM_017026303.1 | XP_016881792.1 | 5 | 1895 | |||