siRNA Details
| Gene Symbol: METTL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136107.1 | NP_001129579.1 | 3 | 498 | ||
| XM_011509232.2 | XP_011507534.1 | 4 | 519 | |||
| XM_011509233.2 | XP_011507535.1 | 5 | 485 | |||
| XM_011509234.2 | XP_011507536.1 | 4 | 497 | |||
| GenBank mRNA | BC157860.1 | - | 3 | 498 | 9057079 | |
| BC171858.1 | - | 3 | 498 | 9054512 | ||

METTL11B
Genomic Map