siRNA Details
| Gene Symbol: FAR2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046258.1 | 6 | 2030 | |||
| NR_046259.1 | 5 | 1932 | ||||
| NR_046260.1 | 4 | 1807 | ||||
| GenBank mRNA | AK054990.1 | - | 4 | 1247 | ||
| BC131487.1 | - | 2 | 3855 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAR2P2| Gene Symbol: FAR2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046258.1 | 6 | 2030 | |||
| NR_046259.1 | 5 | 1932 | ||||
| NR_046260.1 | 4 | 1807 | ||||
| GenBank mRNA | AK054990.1 | - | 4 | 1247 | ||
| BC131487.1 | - | 2 | 3855 | |||