siRNA Details
| Gene Symbol: FBXO41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080410.2 | NP_001073879.2 | 12 | 5727 | ||
| XM_005264158.3 | XP_005264215.1 | 10 | 4646 | |||
| XM_006711950.2 | XP_006712013.1 | 17 | 6711 | |||
| XM_011532562.1 | XP_011530864.1 | 13 | 6159 | |||
| XM_011532563.2 | XP_011530865.1 | 11 | 5084 | |||
| XM_017003421.1 | XP_016858910.1 | 14 | 8808 | |||
| XM_017003422.1 | XP_016858911.1 | 17 | 8453 | |||
| XM_017003423.1 | XP_016858912.1 | 17 | 7834 | |||
| GenBank mRNA | AB075820.1 | - | 12 | 5565 | ||
| AK055121.1 | - | 1 | 2733 | |||
| AK095028.1 | - | 3 | 3398 | |||

FBXO41
Genomic Map