siRNA Details
| Gene Symbol: LINC01107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037809.1 | 3 | 2190 | |||
| GenBank mRNA | AK057376.1 | - | 3 | 1746 | ||
| BC101453.1 | - | 1 | 507 | |||
| BC101454.1 | - | 1 | 507 | |||
| BC101455.1 | - | 1 | 507 | |||
| BC101456.1 | - | 1 | 507 | |||

LINC01107
Genomic Map