siRNA Details
| Gene Symbol: LOC100505841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195535.1 | NP_001182464.1 | 2 | 179 | ||
| XM_011543086.2 | XP_011541388.1 | 5 | 498 | |||
| GenBank mRNA | BU567915.1 | - | 2 | 182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505841| Gene Symbol: LOC100505841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195535.1 | NP_001182464.1 | 2 | 179 | ||
| XM_011543086.2 | XP_011541388.1 | 5 | 498 | |||
| GenBank mRNA | BU567915.1 | - | 2 | 182 | ||