siRNA Details
| Gene Symbol: LINC00032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026679.1 | 15 | 2628 | |||
| GenBank mRNA | AF418573.2 | - | 15 | 2628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00032| Gene Symbol: LINC00032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026679.1 | 15 | 2628 | |||
| GenBank mRNA | AF418573.2 | - | 15 | 2628 | ||