siRNA Details
| Gene Symbol: CASP16P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132322.1 | 9 | 1025 | |||
| GenBank mRNA | AF098666.1 | - | 9 | 1073 | ||
| BC111724.1 | - | 9 | 1025 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASP16P| Gene Symbol: CASP16P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132322.1 | 9 | 1025 | |||
| GenBank mRNA | AF098666.1 | - | 9 | 1073 | ||
| BC111724.1 | - | 9 | 1025 | |||