siRNA Details
| Gene Symbol: TET3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287491.1 | NP_001274420.1 | 3 | 661 | ||
| XM_005264187.3 | XP_005264244.1 | 4 | 1181 | |||
| XM_011532682.2 | XP_011530984.1 | 4 | 1181 | |||
| XM_011532683.2 | XP_011530985.1 | 4 | 1490 | |||
| XM_011532684.2 | XP_011530986.1 | 4 | 2922 | |||
| XM_011532685.2 | XP_011530987.1 | 3 | 1124 | |||
| XM_011532686.2 | XP_011530988.1 | 3 | 1124 | |||
| XM_011532687.2 | XP_011530989.1 | 3 | 463 | |||
| XM_011532688.2 | XP_011530990.1 | 4 | 1181 | |||
| XM_017003566.1 | XP_016859055.1 | 4 | 3021 | |||
| XM_017003567.1 | XP_016859056.1 | 3 | 493 | |||
| GenBank mRNA | HQ220209.1 | - | 3 | 661 | ||

TET3
Genomic Map