siRNA Details
| Gene Symbol: SERPINB9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033851.1 | 2 | 1000 | |||
| GenBank mRNA | BC025340.1 | - | 2 | 1000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINB9P1| Gene Symbol: SERPINB9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033851.1 | 2 | 1000 | |||
| GenBank mRNA | BC025340.1 | - | 2 | 1000 | ||