siRNA Details
| Gene Symbol: FRYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015030.1 | NP_055845.1 | 4 | 669 | ||
| XM_005248090.4 | XP_005248147.1 | 4 | 682 | |||
| XM_011513680.2 | XP_011511982.1 | 1 | 144 | |||
| XM_011513681.2 | XP_011511983.1 | 1 | 144 | |||
| XM_011513682.2 | XP_011511984.1 | 1 | 144 | |||
| XM_011513683.2 | XP_011511985.1 | 3 | 518 | |||
| XM_011513685.2 | XP_011511987.1 | 1 | 144 | |||
| XM_017008046.1 | XP_016863535.1 | 5 | 994 | |||
| GenBank mRNA | AK131250.1 | - | 2 | 344 | ||
| AL833444.1 | - | 4 | 629 | |||

FRYL
Genomic Map