siRNA Details
| Gene Symbol: ENDOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015036.2 | NP_055851.1 | 2 | 3026 | ||
| GenBank mRNA | AB020637.1 | - | 2 | 2979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENDOD1| Gene Symbol: ENDOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015036.2 | NP_055851.1 | 2 | 3026 | ||
| GenBank mRNA | AB020637.1 | - | 2 | 2979 | ||