siRNA Details
| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015174.1 | NP_055989.1 | 19 | 2926 | ||
| XM_011527831.2 | XP_011526133.1 | 19 | 2928 | |||
| GenBank mRNA | AB029009.1 | - | 18 | 2804 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFR2| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015174.1 | NP_055989.1 | 19 | 2926 | ||
| XM_011527831.2 | XP_011526133.1 | 19 | 2928 | |||
| GenBank mRNA | AB029009.1 | - | 18 | 2804 | ||