siRNA Details
| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015174.1 | NP_055989.1 | 19 | 4645 | ||
| GenBank mRNA | AB029009.1 | - | 18 | 4523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFR2| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015174.1 | NP_055989.1 | 19 | 4645 | ||
| GenBank mRNA | AB029009.1 | - | 18 | 4523 | ||