siRNA Details
| Gene Symbol: NBEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015175.2 | NP_055990.1 | 21 | 3250 | ||
| XM_006713072.3 | XP_006713135.1 | 20 | 3166 | |||
| XM_011533533.2 | XP_011531835.1 | 21 | 3247 | |||
| XM_017006010.1 | XP_016861499.1 | 21 | 3247 | |||
| XM_017006011.1 | XP_016861500.1 | 21 | 3103 | |||
| XM_017006012.1 | XP_016861501.1 | 20 | 3166 | |||
| XM_017006013.1 | XP_016861502.1 | 21 | 3247 | |||
| XM_017006014.1 | XP_016861503.1 | 20 | 3022 | |||
| XM_017006015.1 | XP_016861504.1 | 19 | 2878 | |||
| XM_017006016.1 | XP_016861505.1 | 16 | 3824 | |||
| GenBank mRNA | AB011112.3 | - | 9 | 1486 | ||
| AK131104.1 | - | 21 | 3090 | |||

NBEAL2
Genomic Map