siRNA Details
| Gene Symbol: FKBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015258.1 | NP_056073.1 | 3 | 303 | ||
| XM_006717018.2 | XP_006717081.1 | 3 | 220 | |||
| XM_006717019.1 | XP_006717082.1 | 2 | 151 | |||
| XM_017014552.1 | XP_016870041.1 | 2 | 151 | |||
| GenBank mRNA | AB014574.1 | - | 3 | 230 | ||
| AK303074.1 | - | 2 | 151 | |||
| AK307507.1 | - | 7 | 737 | |||
| BC009609.1 | - | 3 | 260 | |||
| DC321217.1 | - | 3 | 304 | |||

FKBP15
Genomic Map