siRNA Details
| Gene Symbol: PHF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304333.2 | NP_001291262.1 | 8 | 3131 | ||
| NM_015297.2 | NP_056112.1 | 8 | 3127 | |||
| NR_136307.1 | 8 | 3131 | ||||
| NR_136308.1 | 8 | 3127 | ||||
| XM_011517827.1 | XP_011516129.1 | 9 | 3119 | |||
| XM_011517828.2 | XP_011516130.1 | 8 | 3223 | |||
| XM_011517829.1 | XP_011516131.1 | 8 | 3381 | |||
| XM_017014553.1 | XP_016870042.1 | 10 | 3413 | |||
| XM_017014554.1 | XP_016870043.1 | 10 | 3193 | |||
| XM_017014555.1 | XP_016870044.1 | 10 | 3165 | |||
| XM_017014556.1 | XP_016870045.1 | 10 | 3697 | |||
| GenBank mRNA | AB028968.1 | - | 8 | 2993 | ||

PHF24
Genomic Map