siRNA Details
| Gene Symbol: HAUS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015302.1 | NP_056117.1 | 19 | 2612 | ||
| XM_011526684.2 | XP_011524986.1 | 18 | 2499 | |||
| GenBank mRNA | AB020648.1 | - | 19 | 2585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HAUS5| Gene Symbol: HAUS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015302.1 | NP_056117.1 | 19 | 2612 | ||
| XM_011526684.2 | XP_011524986.1 | 18 | 2499 | |||
| GenBank mRNA | AB020648.1 | - | 19 | 2585 | ||