siRNA Details
| Gene Symbol: KIAA0368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080398.1 | NP_001073867.1 | 7 | 996 | ||
| XM_005251850.3 | XP_005251907.1 | 5 | 499 | |||
| XM_005251851.4 | XP_005251908.1 | 6 | 878 | |||
| XM_005251852.4 | XP_005251909.1 | 6 | 830 | |||
| XM_005251853.3 | XP_005251910.1 | 6 | 612 | |||
| XM_005251854.3 | XP_005251911.1 | 5 | 781 | |||
| XM_011518422.2 | XP_011516724.1 | 5 | 499 | |||
| GenBank mRNA | AK122712.1 | - | 5 | 483 | ||
| AK308313.1 | - | 6 | 603 | |||

KIAA0368
Genomic Map