siRNA Details
Gene Symbol: TTC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015351.1 | NP_056166.1 | 3 | 1952 | ||
GenBank mRNA | BC047950.1 | - | 3 | 1774 | ||
D86980.1 | - | 3 | 1952 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TTC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015351.1 | NP_056166.1 | 3 | 1952 | ||
GenBank mRNA | BC047950.1 | - | 3 | 1774 | ||
D86980.1 | - | 3 | 1952 |