siRNA Details
| Gene Symbol: ZNF503-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038223.1 | 3 | 1685 | |||
| NR_038224.1 | 3 | 1343 | ||||
| NR_038225.1 | 4 | 1454 | ||||
| NR_038226.1 | 4 | 1466 | ||||
| GenBank mRNA | AK023445.1 | - | 3 | 1678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF503-AS1| Gene Symbol: ZNF503-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038223.1 | 3 | 1685 | |||
| NR_038224.1 | 3 | 1343 | ||||
| NR_038225.1 | 4 | 1454 | ||||
| NR_038226.1 | 4 | 1466 | ||||
| GenBank mRNA | AK023445.1 | - | 3 | 1678 | ||