siRNA Details
| Gene Symbol: EIF4E1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099408.1 | NP_001092878.1 | 9 | 1290 | ||
| XM_011534499.2 | XP_011532801.1 | 9 | 1371 | |||
| GenBank mRNA | AK094270.1 | - | 6 | 2140 | ||
| BM709255.1 | - | 4 | 378 | |||

EIF4E1B
Genomic Map