siRNA Details
| Gene Symbol: TTC41P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027249.1 | 2 | 618 | |||
| GenBank mRNA | AK299490.1 | - | 2 | 538 | ||
| BC016335.1 | - | 2 | 450 | |||
| BC033128.1 | - | 6 | 1199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC41P| Gene Symbol: TTC41P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027249.1 | 2 | 618 | |||
| GenBank mRNA | AK299490.1 | - | 2 | 538 | ||
| BC016335.1 | - | 2 | 450 | |||
| BC033128.1 | - | 6 | 1199 | |||