siRNA Details
| Gene Symbol: SERPINA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080451.1 | NP_001073920.1 | 2 | 222 | ||
| GenBank mRNA | BC146962.1 | - | 2 | 186 | 9056890 | |
| BX248259.1 | - | 2 | 218 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINA11| Gene Symbol: SERPINA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080451.1 | NP_001073920.1 | 2 | 222 | ||
| GenBank mRNA | BC146962.1 | - | 2 | 186 | 9056890 | |
| BX248259.1 | - | 2 | 218 | |||