siRNA Details
| Gene Symbol: HSPA12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025015.2 | NP_079291.2 | 5 | 585 | ||
| XM_005269672.3 | XP_005269729.1 | 6 | 735 | |||
| XM_005269673.4 | XP_005269730.1 | 6 | 1679 | |||
| XM_011539579.2 | XP_011537881.1 | 7 | 796 | |||
| XM_011539580.2 | XP_011537882.1 | 6 | 741 | |||
| GenBank mRNA | AB007877.1 | - | 5 | 585 | ||
| AK294672.1 | - | 4 | 399 | |||

HSPA12A
Genomic Map