siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 10 | 1002 | ||
| XM_005260684.3 | XP_005260741.1 | 10 | 999 | |||
| XM_017027783.1 | XP_016883272.1 | 7 | 623 | |||
| GenBank mRNA | AK226100.1 | - | 5 | 426 | ||
| AK303375.1 | - | 1 | 374 | |||
| AK310540.1 | - | 10 | 1020 | |||
| BC122529.1 | - | 1 | 220 | 40012183 | ||
| BX424937.2 | - | 10 | 948 | |||

C20orf194
Genomic Map