siRNA Details
| Gene Symbol: TMEM251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098621.1 | NP_001092091.1 | 2 | 544 | ||
| NM_015676.1 | NP_056491.1 | 2 | 170 | |||
| GenBank mRNA | AL080118.1 | - | 2 | 139 | ||
| AW404266.1 | - | 2 | 176 | |||
| BC030119.2 | - | 2 | 137 | |||
| CX785861.1 | - | 2 | 544 | |||

TMEM251
Genomic Map