siRNA Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 5 | 432 | |||
| NR_024429.1 | 5 | 528 | ||||
| NR_024431.2 | 7 | 724 | ||||
| GenBank mRNA | AK097923.1 | - | 1 | 441 | ||
| AK098249.1 | - | 7 | 1169 | |||
| BC102000.1 | - | 4 | 275 | |||
| BC102001.1 | - | 4 | 275 | |||
| BC105703.1 | - | 4 | 371 | |||

ZMIZ1-AS1
Genomic Map