siRNA Details
| Gene Symbol: TRIM64C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206631.1 | NP_001193560.1 | 6 | 978 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM64C| Gene Symbol: TRIM64C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206631.1 | NP_001193560.1 | 6 | 978 | ||