siRNA Details
| Gene Symbol: DDX43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018665.2 | NP_061135.2 | 7 | 1223 | ||
| XM_011535926.1 | XP_011534228.1 | 6 | 1091 | |||
| XM_011535927.2 | XP_011534229.1 | 6 | 680 | |||
| XM_011535928.2 | XP_011534230.1 | 6 | 869 | |||
| XM_017011003.1 | XP_016866492.1 | 6 | 809 | |||
| GenBank mRNA | AJ278110.1 | - | 7 | 1086 | ||
| AK303324.1 | - | 6 | 850 | |||
| AK313686.1 | - | 7 | 923 | |||
| AL136751.1 | - | 7 | 911 | |||
| BC066938.1 | - | 7 | 904 | 4829072 | ||

DDX43
Genomic Map