siRNA Details
| Gene Symbol: GXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099650.1 | NP_001093120.1 | 2 | 562 | ||
| NM_173601.1 | NP_775872.1 | 3 | 655 | |||
| XM_017019211.1 | XP_016874700.1 | 3 | 394 | |||
| GenBank mRNA | AK125141.1 | - | 2 | 415 | ||
| BC030023.1 | - | 3 | 663 | 5266607 | ||
| BC039145.1 | - | 2 | 402 | 4508300 | ||

GXYLT1
Genomic Map