siRNA Details
Gene Symbol: LOC283710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243538.1 | NP_001230467.1 | 4 | 491 | ||
XM_011521463.2 | XP_011519765.1 | 4 | 539 | |||
XM_011521465.2 | XP_011519767.1 | 4 | 490 | |||
XM_017022084.1 | XP_016877573.1 | 3 | 624 | |||
GenBank mRNA | BF974240.1 | - | 4 | 489 | ||
BQ055725.1 | - | 3 | 370 |
