siRNA Details
| Gene Symbol: LOC283710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243538.1 | NP_001230467.1 | 4 | 917 | ||
| XM_011521465.2 | XP_011519767.1 | 5 | 774 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283710| Gene Symbol: LOC283710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243538.1 | NP_001230467.1 | 4 | 917 | ||
| XM_011521465.2 | XP_011519767.1 | 5 | 774 | |||