siRNA Details
Gene Symbol: ZNF843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136509.1 | NP_001129981.1 | 2 | 1833 | ||
GenBank mRNA | AK291063.1 | - | 2 | 1591 | ||
BC036762.1 | - | 2 | 1459 | 5588928 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136509.1 | NP_001129981.1 | 2 | 1833 | ||
GenBank mRNA | AK291063.1 | - | 2 | 1591 | ||
BC036762.1 | - | 2 | 1459 | 5588928 |