siRNA Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204210.1 | NP_001191139.1 | 5 | 1066 | ||
| NM_001204211.1 | NP_001191140.1 | 4 | 985 | |||
| NM_001204212.1 | NP_001191141.1 | 3 | 847 | |||
| XM_011524620.2 | XP_011522922.1 | 6 | 604 | |||
| XM_011524624.2 | XP_011522926.1 | 4 | 442 | |||
| XM_017024459.1 | XP_016879948.1 | 5 | 517 | |||
| XM_017024460.1 | XP_016879949.1 | 6 | 604 | |||
| GenBank mRNA | BC042066.1 | - | 3 | 847 | ||
| BC148207.2 | - | 3 | 674 | |||
| BI757060.1 | - | 4 | 404 | |||
| BI757981.1 | - | 4 | 336 | |||

TMEM235
Genomic Map