siRNA Details
| Gene Symbol: C17orf105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136483.1 | NP_001129955.1 | 3, 4 | 350 | ||
| XM_011524633.1 | XP_011522935.1 | 4, 5 | 1589 | |||
| GenBank mRNA | AK125817.1 | - | 3, 4 | 325 | ||
| BC054872.1 | - | 3, 4 | 354 | |||
| BC070324.1 | - | 3, 4 | 350 | |||
| BC146971.1 | - | 3, 4 | 349 | 9056899 | ||

C17orf105
Genomic Map