siRNA Details
| Gene Symbol: LINC00908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015417.1 | 3 | 1724 | |||
| GenBank mRNA | AK093936.1 | - | 3 | 1724 | ||
| BC121150.1 | - | 1 | 1181 | |||
| BC121151.1 | - | 1 | 1181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00908| Gene Symbol: LINC00908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015417.1 | 3 | 1724 | |||
| GenBank mRNA | AK093936.1 | - | 3 | 1724 | ||
| BC121150.1 | - | 1 | 1181 | |||
| BC121151.1 | - | 1 | 1181 | |||