siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 6 | 1637 | |||
| NR_047521.1 | 5 | 1484 | ||||
| NR_047523.1 | 4 | 1388 | ||||
| NR_047524.1 | 4 | 1300 | ||||
| NR_047525.1 | 5 | 1351 | ||||
| GenBank mRNA | AK026873.1 | - | 7 | 1235 | ||
| AK311732.1 | - | 6 | 1338 | |||
| BC042880.1 | - | 5 | 1466 | |||

LINC01128
Genomic Map