siRNA Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122962.1 | NP_001116434.1 | 2 | 622 | ||
| XM_005260708.4 | XP_005260765.1 | 2 | 431 | |||
| XM_011529221.2 | XP_011527523.1 | 2 | 1218 | |||
| XM_011529223.2 | XP_011527525.1 | 3 | 666 | |||
| XM_011529224.2 | XP_011527526.1 | 2 | 110 | |||
| GenBank mRNA | AK095499.1 | - | 2 | 431 | ||
| AK294440.1 | - | 3 | 317 | |||
| BM285394.1 | - | 2 | 624 | |||
| BQ635335.1 | - | 2 | 691 | |||
| DC313096.1 | - | 2 | 520 | |||
| DC358609.1 | - | 2 | 485 | |||

SIRPB2
Genomic Map