siRNA Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122962.1 | NP_001116434.1 | 5 | 2470 | ||
| NM_001134836.1 | NP_001128308.1 | 5 | 2176 | |||
| GenBank mRNA | AK095499.1 | - | 5 | 2275 | ||
| BG539998.1 | - | 1 | 630 | |||

SIRPB2
Genomic Map