siRNA Details
| Gene Symbol: LOC100652871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530666.1 | XP_011528968.1 | 4 | 518 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100652871| Gene Symbol: LOC100652871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530666.1 | XP_011528968.1 | 4 | 518 | ||