siRNA Details
| Gene Symbol: LOC285095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135766.1 | 2 | 140 | |||
| NR_135767.1 | 2 | 100 | ||||
| NR_135768.1 | 2 | 39 | ||||
| GenBank mRNA | AA250751.1 | - | 2 | 66 | ||
| BX108328.1 | - | 2 | 106 | |||
| HY129633.1 | - | 2 | 141 | |||

LOC285095
Genomic Map