siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289947.1 | NP_001276876.1 | 1, 2 | 80 | ||
| GenBank mRNA | BC127676.1 | - | 1, 2 | 80 | 40127368 | |
| BC150546.1 | - | 1, 2 | 80 | |||
| BQ429225.1 | - | 78 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH2| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289947.1 | NP_001276876.1 | 1, 2 | 80 | ||
| GenBank mRNA | BC127676.1 | - | 1, 2 | 80 | 40127368 | |
| BC150546.1 | - | 1, 2 | 80 | |||
| BQ429225.1 | - | 78 | ||||